siRNA Details
| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271212.1 | NP_001258141.1 | 14 | 1504 | ||
| GenBank mRNA | FQ217242.1 | - | 1506 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Atg13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271212.1 | NP_001258141.1 | 14 | 1504 | ||
| GenBank mRNA | FQ217242.1 | - | 1506 | |||
Atg13
Genomic Map