siRNA Details
| Gene Symbol: Trpc4ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100748.1 | NP_001094218.1 | 16 | 1943 | ||
| GenBank mRNA | AY724540.1 | - | 16 | 1915 | ||
| BC087662.1 | - | 16 | 1896 | |||
| BC105861.1 | - | 5 | 442 | |||
| BC166758.1 | - | 16 | 1916 | 7930596 | ||

Trpc4ap
Genomic Map