siRNA Details
| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108690.1 | NP_001102160.1 | 15 | 1735 | ||
| XM_006239245.2 | XP_006239307.1 | 16 | 1766 | |||
| GenBank mRNA | FQ216974.1 | - | 16 | 1725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emc1| Gene Symbol: Emc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108690.1 | NP_001102160.1 | 15 | 1735 | ||
| XM_006239245.2 | XP_006239307.1 | 16 | 1766 | |||
| GenBank mRNA | FQ216974.1 | - | 16 | 1725 | ||