siRNA Details
| Gene Symbol: Vwa3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244859.2 | XP_006244921.1 | 15 | 2760 | ||
| XM_006244860.2 | XP_006244922.1 | 15 | 2761 | |||
| XM_008767048.1 | XP_008765270.1 | 16 | 2325 | |||
| XM_008767049.1 | XP_008765271.1 | 15 | 2760 | |||
| XM_008767050.1 | XP_008765272.1 | 15 | 2759 | |||
| XM_008767051.1 | XP_008765273.1 | 15 | 2762 | |||

Vwa3b
Genomic Map