siRNA Details
| Gene Symbol: Skiv2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034093.1 | NP_001029265.1 | 21 | 2594 | ||
| GenBank mRNA | BC091305.1 | - | 3 | 235 | ||
| CF977889.1 | - | 7 | 710 | |||
| CO401332.1 | - | 3 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Skiv2l2| Gene Symbol: Skiv2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034093.1 | NP_001029265.1 | 21 | 2594 | ||
| GenBank mRNA | BC091305.1 | - | 3 | 235 | ||
| CF977889.1 | - | 7 | 710 | |||
| CO401332.1 | - | 3 | 234 | |||