siRNA Details
| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108977.1 | NP_001102447.1 | 5 | 1324 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha2| Gene Symbol: Epha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108977.1 | NP_001102447.1 | 5 | 1324 | ||