siRNA Details
| Gene Symbol: Ano7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004071.1 | NP_001004071.1 | 8 | 784 | ||
| XM_008767382.1 | XP_008765604.1 | 9 | 2438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano7| Gene Symbol: Ano7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004071.1 | NP_001004071.1 | 8 | 784 | ||
| XM_008767382.1 | XP_008765604.1 | 9 | 2438 | |||