siRNA Details
| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 31 | 5365 | ||
| GenBank mRNA | M76767.1 | - | 31 | 5365 | ||
| X13415.1 | - | 29 | 5163 | |||
| X62888.1 | - | 31 | 5370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fasn| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 31 | 5365 | ||
| GenBank mRNA | M76767.1 | - | 31 | 5365 | ||
| X13415.1 | - | 29 | 5163 | |||
| X62888.1 | - | 31 | 5370 | |||