siRNA Details
| Gene Symbol: Cltc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019299.1 | NP_062172.1 | 29 | 4730 | ||
| XM_006247107.2 | XP_006247169.1 | 30 | 5042 | |||
| XM_006247108.2 | XP_006247170.1 | 29 | 5038 | |||
| XM_006247109.2 | XP_006247171.1 | 30 | 5041 | |||
| XM_008768108.1 | XP_008766330.1 | 28 | 5257 | |||
| GenBank mRNA | J03583.1 | - | 29 | 4730 | ||

Cltc
Genomic Map