siRNA Details
| Gene Symbol: Cd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021744.1 | NP_068512.1 | 2 | 1105 | ||
| XM_006254603.2 | XP_006254665.1 | 2 | 1437 | |||
| GenBank mRNA | AF087943.1 | - | 2 | 1105 | ||
| BC061733.1 | - | 2 | 1243 | 5598794 | ||
| FQ219480.1 | - | 2 | 1138 | |||
| U51804.1 | - | 2 | 1124 | |||

Cd14
Genomic Map