siRNA Details
| Gene Symbol: Syt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053325.1 | NP_445777.1 | 2 | 226 | ||
| GenBank mRNA | AF375462.1 | - | 2 | 226 | ||
| AF375467.1 | - | 2 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syt8| Gene Symbol: Syt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053325.1 | NP_445777.1 | 2 | 226 | ||
| GenBank mRNA | AF375462.1 | - | 2 | 226 | ||
| AF375467.1 | - | 2 | 226 | |||