siRNA Details
| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022608.1 | NP_072130.1 | 12 | 1189 | ||
| GenBank mRNA | AJ132352.1 | - | 12 | 1204 | ||
| BC162038.1 | - | 12 | 1189 | 7099625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec14l3| Gene Symbol: Sec14l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022608.1 | NP_072130.1 | 12 | 1189 | ||
| GenBank mRNA | AJ132352.1 | - | 12 | 1204 | ||
| BC162038.1 | - | 12 | 1189 | 7099625 | ||