siRNA Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032462.2 | NP_115851.1 | 5 | 435 | ||
| XM_006234989.2 | XP_006235051.1 | 5 | 564 | |||
| XM_006234990.2 | XP_006235052.1 | 4 | 3880 | |||
| GenBank mRNA | AB043892.1 | - | 4 | 414 | ||
| AF297118.1 | - | 5 | 437 | |||

Kcnip3
Genomic Map