siRNA Details
| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031780.1 | NP_113968.1 | 11 | 2030 | ||
| GenBank mRNA | AF029107.1 | - | 11 | 2030 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apba2| Gene Symbol: Apba2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031780.1 | NP_113968.1 | 11 | 2030 | ||
| GenBank mRNA | AF029107.1 | - | 11 | 2030 | ||