siRNA Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031813.1 | NP_114001.1 | 4 | 576 | ||
| XM_006249888.2 | XP_006249950.1 | 4 | 686 | |||
| XM_006249889.2 | XP_006249951.1 | 4 | 685 | |||
| GenBank mRNA | AF077338.1 | - | 4 | 576 | ||
| BC061993.1 | - | 4 | 615 | 5599482 | ||

Mybph
Genomic Map