siRNA Details
| Gene Symbol: Rab3il1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134411.1 | NP_599238.1 | 5 | 677 | ||
| GenBank mRNA | AY026049.1 | - | 5 | 677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab3il1| Gene Symbol: Rab3il1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134411.1 | NP_599238.1 | 5 | 677 | ||
| GenBank mRNA | AY026049.1 | - | 5 | 677 | ||