This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AKR1C14 SIRNA Assays ›
Gene Symbol:
Akr1c14
Gene Name:
aldo-keto reductase family 1, member C14
Gene Aliases:
Akr1c9
Chromosome Location:
Chr.17: 69806095 - 69827110 on Build Rnor_6.0
UniGene ID:
Rn.10021
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS332230
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Akr1c14
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138547.2 NP_612556.1 7 852
GenBank mRNA BC091123.1 - 7 852 7372929
D17310.1 - 7 1040
M64393.1 - 7 851

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

steroid metabolic process
hippocampus development
oxidation-reduction process
steroid dehydrogenase activity
androsterone dehydrogenase (B-specific) activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-wnzdn:80/100.66.79.155:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline