siRNA Details
| Gene Symbol: Ctsj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017121.1 | NP_058817.1 | 6, 7 | 859 | ||
| XM_008771439.1 | XP_008769661.1 | 5, 6 | 774 | |||
| GenBank mRNA | AF310623.2 | - | 6, 7 | 859 | ||
| BC097263.1 | - | 6, 7 | 842 | 7460052 | ||
| L14776.1 | - | 3, 4 | 485 | |||

Ctsj
Genomic Map