siRNA Details
| Gene Symbol: Fdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017126.1 | NP_058822.1 | 2 | 369 | ||
| GenBank mRNA | D50436.1 | - | 2 | 369 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fdx1| Gene Symbol: Fdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017126.1 | NP_058822.1 | 2 | 369 | ||
| GenBank mRNA | D50436.1 | - | 2 | 369 | ||