siRNA Details
| Gene Symbol: Eif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106867.1 | NP_001100337.1 | 3 | 397 | ||
| GenBank mRNA | BC169125.1 | - | 3 | 317 | 6919533 | |
| FQ211778.1 | - | 3 | 400 | |||
| FQ214080.1 | - | 3 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif1b| Gene Symbol: Eif1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106867.1 | NP_001100337.1 | 3 | 397 | ||
| GenBank mRNA | BC169125.1 | - | 3 | 317 | 6919533 | |
| FQ211778.1 | - | 3 | 400 | |||
| FQ214080.1 | - | 3 | 410 | |||