siRNA Details
| Gene Symbol: Snx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253168.1 | XP_006253230.1 | 4 | 590 | ||
| XM_224863.6 | XP_224863.5 | 4 | 590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx25| Gene Symbol: Snx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253168.1 | XP_006253230.1 | 4 | 590 | ||
| XM_224863.6 | XP_224863.5 | 4 | 590 | |||