siRNA Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277232.1 | NP_001264161.1 | 8, 9 | 788 | ||
| NM_182953.2 | NP_891998.1 | 8, 9 | 840 | |||
| XM_006234100.1 | XP_006234162.1 | 8, 9 | 863 | |||
| GenBank mRNA | AY334013.1 | - | 7, 8 | 737 | ||
| BC160820.1 | - | 7, 8 | 864 | 7459041 | ||

Nek6
Genomic Map