siRNA Details
| Gene Symbol: Olr964 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001073.1 | NP_001001073.1 | 1 | 462 | ||
| Gene Symbol: LOC100910607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240817.1 | XP_006240879.1 | 2 | 240 | ||

Olr964
Genomic Map