siRNA Details
| Gene Symbol: Fez2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053600.2 | NP_446052.2 | 7 | 1040 | ||
| XM_006239637.2 | XP_006239699.1 | 6 | 1100 | |||
| XM_006239638.2 | XP_006239700.1 | 6 | 1010 | |||
| GenBank mRNA | AB076183.1 | - | 7 | 1040 | ||
| AB080086.1 | - | 6 | 938 | |||
| AF120109.1 | - | 2 | 133 | |||
| AF120110.1 | - | 1 | 52 | |||
| AF120111.1 | - | 1 | 52 | |||
| BC100060.1 | - | 6 | 940 | 7128839 | ||
| CB725415.1 | - | 3 | 282 | |||
| U64689.1 | - | 7 | 866 | |||

Fez2
Genomic Map