siRNA Details
| Gene Symbol: Epn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057136.1 | NP_476477.1 | 3 | 478 | ||
| GenBank mRNA | AF018261.1 | - | 3 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epn1| Gene Symbol: Epn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057136.1 | NP_476477.1 | 3 | 478 | ||
| GenBank mRNA | AF018261.1 | - | 3 | 478 | ||