siRNA Details
| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013098.2 | NP_037230.2 | 1 | 146 | ||
| GenBank mRNA | BC090067.1 | - | 1 | 146 | 7109006 | |
| D78592.1 | - | 1 | 158 | |||
| L37333.1 | - | 1 | 110 | |||
| U07993.1 | - | 1 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G6pc| Gene Symbol: G6pc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013098.2 | NP_037230.2 | 1 | 146 | ||
| GenBank mRNA | BC090067.1 | - | 1 | 146 | 7109006 | |
| D78592.1 | - | 1 | 158 | |||
| L37333.1 | - | 1 | 110 | |||
| U07993.1 | - | 1 | 108 | |||