This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CLEC2L SIRNA Assays ›
Gene Symbol:
Clec2l
Gene Name:
C-type lectin domain family 2, member L
Gene Aliases:
RGD1565373
Chromosome Location:
Chr.4: 66405166 - 66421654 on Build Rnor_6.0
UniGene ID:
Rn.162737
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS341643
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clec2l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044233.2 NP_001037698.2 3 308
GenBank mRNA BC161812.1 - 438
DQ630549.1 - 3 168

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001101507, CJ059176, CJ059317.

Gene Ontology Categories:

Process(es)

carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qx8c9:80/100.66.76.99:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline