siRNA Details
| Gene Symbol: Acy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005383.1 | NP_001005383.1 | 3 | 213 | ||
| XM_006243715.2 | XP_006243777.1 | 4 | 515 | |||
| XM_006243716.2 | XP_006243778.1 | 4 | 519 | |||
| GenBank mRNA | AY580164.1 | - | 3 | 213 | ||
| BC078930.1 | - | 4 | 276 | 7107165 | ||
| FQ232544.1 | - | 4 | 370 | |||

Acy1
Genomic Map