siRNA Details
| Gene Symbol: Plet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014209.1 | NP_001014231.1 | 2 | 376 | ||
| GenBank mRNA | BC088858.1 | - | 2 | 376 | 7308076 | |
| FQ228964.1 | - | 2 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plet1| Gene Symbol: Plet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014209.1 | NP_001014231.1 | 2 | 376 | ||
| GenBank mRNA | BC088858.1 | - | 2 | 376 | 7308076 | |
| FQ228964.1 | - | 2 | 399 | |||