This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZC4H2 SIRNA Assays ›
Gene Symbol:
Zc4h2
Gene Name:
zinc finger, C4H2 domain containing
Gene Aliases:
RGD1561708
Chromosome Location:
Chr.X: 64887978 - 64908682 on Build Rnor_6.0
UniGene ID:
Rn.12889
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS343239
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price Contact Us
Your Price
Online offer:
3062.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zc4h2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126374.1 NP_001119846.1 3 410
GenBank mRNA BC162002.1 - 3 410 9088857

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001003916, BC079862, BC094942, BC119285, AK162005, and others.

Gene Ontology Categories:

Function(s)

nervous system development

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline