siRNA Details
| Gene Symbol: Slc35a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127642.1 | NP_001121114.1 | 4 | 662 | ||
| XM_006256693.2 | XP_006256755.1 | 4 | 944 | |||
| GenBank mRNA | BC166811.1 | - | 4 | 662 | 7453742 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35a2| Gene Symbol: Slc35a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127642.1 | NP_001121114.1 | 4 | 662 | ||
| XM_006256693.2 | XP_006256755.1 | 4 | 944 | |||
| GenBank mRNA | BC166811.1 | - | 4 | 662 | 7453742 | |