siRNA Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109911.1 | NP_001103381.1 | 15 | 2024 | ||
| NM_053876.2 | NP_446328.2 | 16 | 1907 | |||
| XM_006241116.2 | XP_006241178.1 | 15 | 3214 | |||
| XM_008765198.1 | XP_008763420.1 | 13 | 1754 | |||
| GenBank mRNA | AY724543.1 | - | 15 | 2011 | ||
| BC088317.1 | - | 11 | 1535 | 7325202 | ||
| U32577.2 | - | 16 | 1907 | |||

Hnrnpm
Genomic Map