siRNA Details
| Gene Symbol: Grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133570.5 | NP_598254.2 | 2 | 464 | ||
| GenBank mRNA | AW524119.1 | - | 2 | 416 | ||
| BF567195.1 | - | 2 | 445 | |||
| M31176.1 | - | 2 | 868 | |||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Grp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133570.5 | NP_598254.2 | 2 | 464 | ||
| GenBank mRNA | AW524119.1 | - | 2 | 416 | ||
| BF567195.1 | - | 2 | 445 | |||
| M31176.1 | - | 2 | 868 | |||
Grp
Genomic Map