This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EPO SIRNA Assays ›
Gene Symbol:
Epo
Gene Name:
erythropoietin
Gene Aliases:
-
Chromosome Location:
Chr.12: 22274828 - 22278268 on Build Rnor_6.0
UniGene ID:
Rn.11365
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351246
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Epo
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017001.1 NP_058697.1 3 242
XM_006249101.2 XP_006249163.1 2 384
GenBank mRNA D10763.1 - 3 242
L10608.1 - 2 211

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
aging
response to nutrient
positive regulation of cell proliferation
response to salt stress
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
response to lipopolysaccharide
negative regulation of myeloid cell apoptotic process
response to vitamin A
response to testosterone
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
response to estrogen
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
response to axon injury
response to electrical stimulus
response to hyperoxia
response to interleukin-1
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline