siRNA Details
| Gene Symbol: Abcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022281.2 | NP_071617.2 | 9, 10 | 1265 | ||
| GenBank mRNA | AF487549.1 | - | 9, 10 | 1265 | ||
| AY170916.1 | - | 9, 10 | 1226 | |||
| AY174892.1 | - | 9, 10 | 1265 | |||
| AY174893.1 | - | 9, 10 | 1265 | |||
| Gene Symbol: LOC100912948 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245698.2 | XP_006245760.1 | 3, 4 | 1486 | ||

Abcc1
Genomic Map