siRNA Details
| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113570.1 | NP_001107042.1 | 7 | 836 | ||
| GenBank mRNA | AF391798.1 | - | 7 | 597 | ||
| CB578423.1 | - | 3 | 527 | |||
| CK842762.1 | - | 3 | 509 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf3a| Gene Symbol: Gtf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113570.1 | NP_001107042.1 | 7 | 836 | ||
| GenBank mRNA | AF391798.1 | - | 7 | 597 | ||
| CB578423.1 | - | 3 | 527 | |||
| CK842762.1 | - | 3 | 509 | |||