siRNA Details
| Gene Symbol: Prlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034111.1 | NP_001029283.1 | 8 | 915 | ||
| NM_012630.2 | NP_036762.1 | 8 | 925 | |||
| XM_008760751.1 | XP_008758973.1 | 10 | 2522 | |||
| GenBank mRNA | BC092133.1 | - | 8 | 919 | 7310645 | |
| M19304.1 | - | 8 | 926 | |||
| M34083.1 | - | 7 | 811 | |||
| M57668.1 | - | 8 | 915 | |||
| M74152.1 | - | 8 | 885 | |||
| U07567.1 | - | 8 | 868 | |||

Prlr
Genomic Map