This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAL SIRNA Assays ›
Gene Symbol:
Mal
Gene Name:
mal, T-cell differentiation protein
Gene Aliases:
MALGENE
Chromosome Location:
Chr.3: 120209647 - 120233655 on Build Rnor_6.0
UniGene ID:
Rn.10174
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS351962
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 458.00
Your Price
Online offer:
458.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mal
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012798.1 NP_036930.1 2 232
GenBank mRNA BC085755.1 - 2 193 7108924
U31367.1 - 2 232
X82557.1 - 2 203

Back To Top

More Information


Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
myelination
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
structural constituent of myelin sheath

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-592sj:80/100.66.79.144:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline