This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLR1271 SIRNA Assays ›
Gene Symbol:
Olr1271 LOC100909885 LOC100910120 LOC100910162 LOC100910224 LOC100910294 LOC103693055
Gene Name:
olfactory receptor 1271
olfactory receptor 8D1-like
olfactory receptor 8D1-like
olfactory receptor 8D1-like
olfactory receptor 8D1-like
olfactory receptor 8D1-like
olfactory receptor 8D1-like
Gene Aliases:
Olp4
-
-
-
-
-
-
Chromosome Location:
Chr.8: 42776302 - 42780782 on Build Rnor_6.0
Chromosome Location:
Chr.2: 35178510 - 35179436 on Build Rnor_6.0
Chromosome Location:
Chr.8: 42817144 - 42818067 on Build Rnor_6.0
Chromosome Location:
Chr.2: 115238435 - 115239361 on Build Rnor_6.0
Chromosome Location:
Chr.8: 42846096 - 42847022 on Build Rnor_6.0
Chromosome Location:
Chr.2: 115303322 - 115304248 on Build Rnor_6.0
Chromosome Location:
Chr.8: 42881124 - 42883941 on Build Rnor_6.0
UniGene ID:
Rn.10385
-
-
-
-
-
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
siRNA ID RSS352621
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr1271
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173300.1 NP_775422.1 2 247
GenBank mRNA X80671.1 - 2 247
Gene Symbol: LOC100909885
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749179.2 XP_003749227.2 1 42
Gene Symbol: LOC100910120
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750489.2 XP_003750537.2 1 42
Gene Symbol: LOC100910162
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749263.2 XP_003749311.2 42
Gene Symbol: LOC100910224
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766109.1 XP_008764331.1 1 42
Gene Symbol: LOC100910294
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006232223.1 XP_006232285.1 1 42
Gene Symbol: LOC103693055
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766110.1 XP_008764332.1 42

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity
G-protein coupled receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline