siRNA Details
| Gene Symbol: Kremen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105767.1 | NP_001099237.1 | 8 | 1238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kremen2| Gene Symbol: Kremen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105767.1 | NP_001099237.1 | 8 | 1238 | ||