siRNA Details
| Gene Symbol: Zfp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751129.3 | XP_003751177.1 | 4 | 502 | ||
| XM_221917.8 | XP_221917.5 | 5 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp12| Gene Symbol: Zfp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751129.3 | XP_003751177.1 | 4 | 502 | ||
| XM_221917.8 | XP_221917.5 | 5 | 598 | |||