This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other THSD7B SIRNA Assays ›
Gene Symbol:
Thsd7b
Gene Name:
thrombospondin, type I, domain containing 7B
Gene Aliases:
RGD1306938
Chromosome Location:
Chr.13: 46101061 - 46931619 on Build Rnor_6.0
UniGene ID:
Rn.225282
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS353270
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price 458.00
Your Price
Online offer:
458.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Thsd7b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191669.1 NP_001178598.1 20 3928
XM_008769435.1 XP_008767657.1 21 8316

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001080427, AB051466.
This siRNA also targets the following transcripts in Mouse: NM_172485, BC063250, AK166575.

Panther Classification:

Molecular Function -

cell adhesion molecule

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline