siRNA Details
| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007636.2 | NP_001007637.1 | 2 | 231 | ||
| XM_006232603.2 | XP_006232665.1 | 3 | 915 | |||
| GenBank mRNA | S68809.1 | - | 2 | 201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100a1| Gene Symbol: S100a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007636.2 | NP_001007637.1 | 2 | 231 | ||
| XM_006232603.2 | XP_006232665.1 | 3 | 915 | |||
| GenBank mRNA | S68809.1 | - | 2 | 201 | ||