This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM227B SIRNA Assays ›
Gene Symbol:
Fam227b
Gene Name:
family with sequence similarity 227, member B
Gene Aliases:
RGD1359452
Chromosome Location:
Chr.3: 118268631 - 118427896 on Build Rnor_6.0
UniGene ID:
Rn.146039
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID RSS355507
Size 20 nmol
Availability Made To Order
Catalog # 1330001
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam227b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013920.2 NP_001013942.2 5 413
XM_006234909.2 XP_006234971.1 5 336
XM_008762153.1 XP_008760375.1 5 370
GenBank mRNA BC079426.1 - 5 372 7114265
BC089861.1 - 5 369 7114720
CK653446.1 - 5 469

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cb7vm:80/100.66.79.242:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline