siRNA Details
| Gene Symbol: Fggy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013932.2 | NP_001013954.2 | 9 | 1055 | ||
| XM_006238410.2 | XP_006238472.1 | 9 | 1693 | |||
| XM_006238413.2 | XP_006238475.1 | 10 | 1266 | |||
| XM_008763856.1 | XP_008762078.1 | 9 | 1055 | |||
| XM_008763857.1 | XP_008762079.1 | 4 | 1157 | |||
| XM_008763858.1 | XP_008762080.1 | 5 | 2447 | |||
| XM_008763859.1 | XP_008762081.1 | 5 | 904 | |||
| GenBank mRNA | BC090077.1 | - | 8 | 880 | 7107754 | |

Fggy
Genomic Map