siRNA Details
| Gene Symbol: Nup37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106775.1 | NP_001100245.1 | 2 | 338 | ||
| XM_006241197.1 | XP_006241259.1 | 2 | 243 | |||
| XM_006241198.2 | XP_006241260.1 | 2 | 238 | |||
| GenBank mRNA | BC091253.1 | - | 1 | 100 | ||
| BC161937.1 | - | 2 | 210 | 9022830 | ||
| FQ220712.1 | - | 2 | 221 | |||

Nup37
Genomic Map