siRNA Details
| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135799.1 | NP_001129271.1 | 13 | 2223 | ||
| XM_006242715.2 | XP_006242777.1 | 11 | 3177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmper| Gene Symbol: Bmper | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135799.1 | NP_001129271.1 | 13 | 2223 | ||
| XM_006242715.2 | XP_006242777.1 | 11 | 3177 | |||