siRNA Details
| Gene Symbol: Efcab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013972.2 | NP_001013994.1 | 8 | 1062 | ||
| GenBank mRNA | BC081930.1 | - | 8 | 951 | ||
| BC127522.1 | - | 8 | 1062 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab3| Gene Symbol: Efcab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013972.2 | NP_001013994.1 | 8 | 1062 | ||
| GenBank mRNA | BC081930.1 | - | 8 | 951 | ||
| BC127522.1 | - | 8 | 1062 | |||