siRNA Details
| Gene Symbol: Trim65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135714.1 | NP_001129186.1 | 6 | 1874 | ||
| GenBank mRNA | FQ223610.1 | - | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim65| Gene Symbol: Trim65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135714.1 | NP_001129186.1 | 6 | 1874 | ||
| GenBank mRNA | FQ223610.1 | - | 1445 | |||