siRNA Details
| Gene Symbol: Prr14l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251416.2 | XP_006251478.1 | 5 | 5237 | ||
| XM_008770385.1 | XP_008768607.1 | 5 | 5241 | |||
| XM_008770386.1 | XP_008768608.1 | 5 | 5951 | |||
| XM_008770387.1 | XP_008768609.1 | 5 | 5567 | |||
| XM_008770388.1 | XP_008768610.1 | 4 | 5430 | |||
| XM_223586.7 | XP_223586.5 | 4 | 5078 | |||

Prr14l
Genomic Map